Learn More
Ethyl trifluoroacetate, 99%, Thermo Scientific Chemicals
Description
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl trifluoroacetate is used as an intermediate in organic synthesis to prepare organic fluorine compounds like 3-ethyl-1-methylimidazolium trifluoroacetate (EMITA). It is involved in the syntheses of various pharmaceutically active molecules and agricultural products. It is also useful for the preparation of tri fluoroacylated compounds.
Solubility
Slightly miscible with water. Miscible with chloroform and methanol.
Notes
Moisture Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
Specifications
Melting Point | <-78°C |
Density | 1.192 |
Boiling Point | 60°C to 62°C |
Flash Point | −1°C (30°F) |
Odor | Sweet |
Refractive Index | 1.307 |
Linear Formula | CF3CO2CH2CH3 |
Quantity | 250 g |
UN Number | UN2924 |
Beilstein | 1761411 |
Show More |
Safety and Handling
Hazard Category | H225-H319 |
Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
EINECSNumber | 206-851-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.