Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl trifluoroacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
2.5L
Packaging:
Glass bottle
383-63-1
C4H5F3O2
142.08
MFCD00000419
STSCVKRWJPWALQ-UHFFFAOYSA-N
ethyl trifluoroacetate,trifluoroacetic acid ethyl ester,ethyltrifluoroacetate,acetic acid, trifluoro-, ethyl ester,trifluoroacetic acid, ethyl ester,ethyl trifluoroethanoate,unii-a6tzk6x11x,acetic acid, 2,2,2-trifluoro-, ethyl ester,a6tzk6x11x,ethyl ester of trifluoroacetic acid
9794
ethyl 2,2,2-trifluoroacetate
CCOC(=O)C(F)(F)F
383-63-1
C4H5F3O2
142.08
MFCD00000419
STSCVKRWJPWALQ-UHFFFAOYSA-N
ethyl trifluoroacetate,trifluoroacetic acid ethyl ester,ethyltrifluoroacetate,acetic acid, trifluoro-, ethyl ester,trifluoroacetic acid, ethyl ester,ethyl trifluoroethanoate,unii-a6tzk6x11x,acetic acid, 2,2,2-trifluoro-, ethyl ester,a6tzk6x11x,ethyl ester of trifluoroacetic acid
9794
ethyl 2,2,2-trifluoroacetate
CCOC(=O)C(F)(F)F
Refractive Index 1.3060 to 1.3080
Melting Point -78.0°C
Specific Gravity 1.194
Packaging Glass bottle
Density 1.1940g/mL
Quantity 500mL
Flash Point −1°C
Color Undesignated
Infrared Spectrum Authentic
Boiling Point 60.0°C to 62.0°C
Assay Percent Range 99%
Linear Formula CF3CO2C2H5
Beilstein 02,II,186
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Formula Weight 142.08
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethyl trifluoroacetate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger