Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyltriphenylphosphonium bromide, 98%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1530-32-1
C20H20BrP
371.26
MFCD00011838
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
ethyltriphenylphosphonium bromide,ethyl triphenyl phosphonium bromide,triphenylethylphosphonium bromide,phosphonium, ethyltriphenyl-, bromide,ethyltriphenylphosphanium bromide,ethyl triphenylphosphonium bromide,unii-r85v84ul5v,ethyltriphenylphosphoniumbromide,guaifenesin powder,pubchem18705
73727
ethyl(triphenyl)phosphanium;bromide
CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
1530-32-1
C20H20BrP
371.26
MFCD00011838
JHYNXXDQQHTCHJ-UHFFFAOYSA-M
ethyltriphenylphosphonium bromide,ethyl triphenyl phosphonium bromide,triphenylethylphosphonium bromide,phosphonium, ethyltriphenyl-, bromide,ethyltriphenylphosphanium bromide,ethyl triphenylphosphonium bromide,unii-r85v84ul5v,ethyltriphenylphosphoniumbromide,guaifenesin powder,pubchem18705
73727
ethyl(triphenyl)phosphanium;bromide
CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
for C-C Bond Formation (Olefination)
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Loss on Drying 1% max.
Melting Point 206.5°C to 208.5°C
Quantity 100g
Flash Point >200°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C2H5P(C6H5)3Br
Solubility Information Solubility in water: 120g/L (23°C). Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in arom. hydrocarbons
Formula Weight 371.26
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Ethyltriphenylphosphonium bromide, 99+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-223-3

TSCA : TSCA