Alfa Aesar™ Ethyl undecanoate, 97%

Manufacturer: Alfa Aesar™ B2341614

Quantity 25g

 View more versions of this product

Catalog No. AAB2341614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 627-90-7
    Molecular Formula C13H26O2
    Linear Formula CH3(CH2)9CO2CH2CH3
    Formula Weight 214.35
    MDL Number MFCD00008958
    Beilstein 1767805
    Synonym Undecanoic acid ethyl ester
    Chemical Name or Material Ethyl undecanoate

    Specifications

    Formula Weight 214.35
    TSCA TSCA
    Molecular Formula C13H26O2
    Linear Formula CH3(CH2)9CO2CH2CH3
    MDL Number MFCD00008958
    CAS 627-90-7
    Beilstein 1767805
    Synonym Undecanoic acid ethyl ester
    Flash Point >110°C (230°F)
    Boiling Point 103° to 105°C (4mmHg)
    Melting Point −15°C
    Quantity 25g
    Chemical Name or Material Ethyl undecanoate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit