Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Vinyl Ether, Stabilized, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
109-92-2,1310-58-3
C4H8O
72.11
MFCD00009248
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether,ethoxyethene,ethoxyethylene,ethene, ethoxy,1-ethoxyethene,1-ethoxyethylene,vinyl ethyl ether,vinamar,ether, ethyl vinyl,ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C
109-92-2,1310-58-3
C4H8O
72.11
MFCD00009248
FJKIXWOMBXYWOQ-UHFFFAOYSA-N
ethyl vinyl ether,ethoxyethene,ethoxyethylene,ethene, ethoxy,1-ethoxyethene,1-ethoxyethylene,vinyl ethyl ether,vinamar,ether, ethyl vinyl,ether, vinyl ethyl
8023
ethenoxyethane
CCOC=C
Viscosity 0.28 mPa.s (5°C)
Packaging Glass bottle
Specific Gravity 0.75
Melting Point -116.0°C
Color Colorless
Refractive Index 1.3760 to 1.3780
Boiling Point 36.0°C
Infrared Spectrum Authentic
Density 0.7500g/mL
Stabilizer 0.1% KOH
Quantity 250mL
Flash Point −45°C
Assay Percent Range 99%
Linear Formula C2H5OCH=CH2
Beilstein 01,433
Fieser 01,386; 02,198; 04,234; 10,184; 11,235
Solubility Information Solubility in water: 7.8g/L (25°C). Other solubilities: soluble in most organic solvents
Formula Weight 72.11
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethyl vinyl ether, Stabilized
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger