Alfa Aesar™ Ethylaluminum dichloride, 1M in hexane

Manufacturer: Alfa Aesar™ 41712AD

Quantity 50mL

 View more versions of this product

Catalog No. AA41712AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 563-43-9
    Formula Weight 126.95
    MDL Number MFCD00000457
    UN Number UN3394
    Chemical Name or Material Ethylaluminum dichloride

    Specifications

    Formula Weight 126.95
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Health Hazard 2 GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H318-H336
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.
    RTECS Number BD0705000
    Boiling Point 68° to 70°C
    Sensitivity Air & Moisture Sensitive
    Physical Form Liquid
    MDL Number MFCD00000457
    CAS 563-43-9
    UN Number UN3394
    Flash Point −22° (−9°F)
    Health Hazard 1 Danger
    Density 0.729g/mL
    Color Colorless to pale yellow
    DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
    Concentration 1M in hexane
    TSCA TSCA
    Quantity 50mL
    Chemical Name or Material Ethylaluminum dichloride

    GHS H Statement
    H225-H250-H260-H252-H304-H361-H373-H314-H318-H336
    Highly flammable liquid and vapor.
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gases which may ignite spontaneously.
    Self-heating in large quantities
    may catch fire.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit