Learn More
Ethylaluminum dichloride, 1M in hexanes, Thermo Scientific Chemicals
Description
Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 0.729 g/mL |
Boiling Point | 68°C to 70°C |
Flash Point | -22°C (-9°F) |
Quantity | 10 mL |
UN Number | UN3394 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Soluble in acetone. Miscible with alcohol,chloroform,ether. Reacts with water. |
Formula Weight | 126.95 |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | H225-H250-H260-H304-H314-H335-H336-H361f-H373 |
Hazard Statement | GHS H Statement H225-H250-H260-H252-H304-H361-H373-H314-H318-H336 Highly flammable liquid and vapor. Catches fire spontaneously if exposed to air. In contact with water releases flammable gases which may ignite spontaneously. Self-heating in large quantities may catch fire. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Causes serious eye damage. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P281-P302+P334-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE |
EINECSNumber | 209-248-6 |
RTECSNumber | BD0705000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.