Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene 99.8%, ACROS Organics™

Click to view available options
Quantity:
25mL
500mL
1L
2.5L
25L
Packaging:
Glass bottle
Metal Drum
100-41-4
C8H10
106.17
MFCD00011647
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene
7500
CHEBI:16101
ethylbenzene
CCC1=CC=CC=C1
100-41-4
C8H10
106.17
MFCD00011647
YNQLUTRBYVCPMQ-UHFFFAOYSA-N
phenylethane,ethylbenzol,benzene, ethyl,aethylbenzol,ethylenzene,ethyl benzene,ethylbenzeen,etilbenzene,etylobenzen,alpha-methyltoluene
7500
CHEBI:16101
ethylbenzene
CCC1=CC=CC=C1
Viscosity 0.63 mPa.s (20°C)
Flash Point 22°C
Specific Gravity 0.86
Water 0.02% max. (coulometric)
Density 0.8600g/mL
Quantity 500mL
Color Undesignated
Boiling Point 136.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4940 to 1.4970
Melting Point -95.0°C
Assay Percent Range 99.8%
Linear Formula C6H5C2H5
Beilstein 05, II, 274
Merck Index 15, 3820
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, benzene, carbon tetrachloride,, ether and sulfur dioxide, slightly soluble in chloroform, insoluble in ammonia
Formula Weight 106.17
Physical Form Liquid
Percent Purity 99.8%
Grade Pure
Chemical Name or Material Ethylbenzene, 99.8%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Highly flammable liquid and vapor.

Emergency Overview
Flammable liquid and vapor. Possible cancer hazard. May cause cancer based on animal data. Aspiration hazard if swallowed - can enter lungs and cause damage. Harmful by inhalation. Inhalation may cause central nervous system effects. May be harmful if swallowed or absorbed through the skin. May cause skin, eye, and respiratory tract irritation.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

EINECSNumber : 202-849-4

RTECSNumber : DA0700000

TSCA : TSCA