Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylbenzene Solution, NMR Reference Standard, 0.1% in chloroform-d (99.8 atom% D), TMS 0.01%, MilliporeSigma™ Supelco™

$176.40 - $248.50

Specifications

Grade NMR reference standard
Linear Formula C8H10
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
11-101-8379
View Documents
MilliporeSigma Supelco
4871121EA
0.235 mL 3 mm O.D. tube
Each for $248.50
Add to cart
 
11-101-6587
View Documents
MilliporeSigma Supelco
4871047IN
0.700 mL 5 mm O.D. x 7 in. tube
Each for $176.40
Add to cart
 
11-101-8587
View Documents
MilliporeSigma Supelco
4871048IN
0.700 mL 5 mm O.D. x 8 in. tube
Each for $180.70
Add to cart
 
Description

Description

Environmental
Specifications

Specifications

NMR reference standard
C8H10
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit