Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
9004-57-3
C23H24N6O4
448.48
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
9004-57-3
C23H24N6O4
448.48
ARSKJXYLLONUAJ-UHFFFAOYSA-N
2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

ET110, 9004-57-3

Spectrum™ Ethylcellulose, 10 mPa.s, NF is used as a coating agent, flavoring fixative, tablet binder and filler, film-former, and as a viscosity-increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Chloride 0.001
Ignition Residue 0.005
Percent Purity 44 to 51%
Loss on Drying 0.03
Packaging Fiber Drum
Quantity 12 kg