Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethylcyclopentadienyl)(triphenylphosphine) copper(I), ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
308847-89-4
C25H24CuP
418.987
MFCD02093653
DSONSPGWGUJRLY-UHFFFAOYSA-N
ethylcyclopentadienyl triphenylphosphine copper i,1-ethylcyclopenta-2,4-dien-1-yl copper; triphenylphosphine
71311150
copper(1+);5-ethylcyclopenta-1,3-diene;triphenylphosphane
CC[C-]1C=CC=C1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cu+]
308847-89-4
C25H24CuP
418.987
MFCD02093653
DSONSPGWGUJRLY-UHFFFAOYSA-N
ethylcyclopentadienyl triphenylphosphine copper i,1-ethylcyclopenta-2,4-dien-1-yl copper; triphenylphosphine
71311150
copper(1+);5-ethylcyclopenta-1,3-diene;triphenylphosphane
CC[C-]1C=CC=C1.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cu+]
Quantity 1g
Color Beige
Packaging Glass bottle
Infrared Spectrum Authentic
Formula Weight 418.99
Physical Form Crystalline Powder
Chemical Name or Material (Ethylcyclopentadienyl)(triphenylphosphine) copper(I)

AIR SENSITIVE

MOISTURE SENSITIVE