Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
42303-42-4
C6H11NO2·ClH
165.62
MFCD00190747
XFNUTZWASODOQK-UHFFFAOYSA-N
ethyl 1-aminocyclopropanecarboxylate hydrochloride,1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride,h-acpc-oet.hcl,ethyl 1-aminocyclopropane-1-carboxylate hydrochloride,1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride,h-acpc-oet hcl,1-aminocyclopropane-1-carboxylic acid ethyl ester,acpc-oet-hcl,h-acpc-oet?cl,pubchem14733
386203
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
CCOC(=O)C1(CC1)N.Cl
42303-42-4
C6H11NO2·ClH
165.62
MFCD00190747
XFNUTZWASODOQK-UHFFFAOYSA-N
ethyl 1-aminocyclopropanecarboxylate hydrochloride,1-aminocyclopropane-1-carboxylic acid ethyl ester hydrochloride,h-acpc-oet.hcl,ethyl 1-aminocyclopropane-1-carboxylate hydrochloride,1-amino-cyclopropyl-1-carboxylic acid ethyl ester hydrochloride,h-acpc-oet hcl,1-aminocyclopropane-1-carboxylic acid ethyl ester,acpc-oet-hcl,h-acpc-oet?cl,pubchem14733
386203
ethyl 1-aminocyclopropane-1-carboxylate;hydrochloride
CCOC(=O)C1(CC1)N.Cl
Melting Point 113.0°C to 114.0°C
Quantity 5g
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Elemental Analysis 42.41 to 45.48% C, 8.24 to 8.66% N
Assay Percent Range 98%
Formula Weight 165.62
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Ethyl 1-aminocyclopropanecarboxylate hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning