Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,4-dioxovalerate, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
615-79-2
C7H10O4
158.15
MFCD00009124
OYQVQWIASIXXRT-UHFFFAOYSA-N
ethyl 2,4-dioxovalerate,ethyl acetopyruvate,pentanoic acid, 2,4-dioxo-, ethyl ester,ethyl2,4-dioxovalerate,2,4-dioxo-pentanoic acid ethyl ester,2,4-dioxopentanoic acid ethyl ester,2,4-dioxo-pentanoic-acid-ethyl-ester,2,4-dioxo-pentanoicacid ethylester,ethyl 2,4-dioxo-valerate,ethyl 2,4-dioxo-pentanoate
69208
ethyl 2,4-dioxopentanoate
CCOC(=O)C(=O)CC(=O)C
615-79-2
C7H10O4
158.15
MFCD00009124
OYQVQWIASIXXRT-UHFFFAOYSA-N
ethyl 2,4-dioxovalerate,ethyl acetopyruvate,pentanoic acid, 2,4-dioxo-, ethyl ester,ethyl2,4-dioxovalerate,2,4-dioxo-pentanoic acid ethyl ester,2,4-dioxopentanoic acid ethyl ester,2,4-dioxo-pentanoic-acid-ethyl-ester,2,4-dioxo-pentanoicacid ethylester,ethyl 2,4-dioxo-valerate,ethyl 2,4-dioxo-pentanoate
69208
ethyl 2,4-dioxopentanoate
CCOC(=O)C(=O)CC(=O)C
Specific Gravity 1.12
Refractive Index 1.4730 to 1.4750
Quantity 25g
Infrared Spectrum Authentic
Flash Point >112°C
Boiling Point 101.0°C to 103.0°C (12.0 mmHg)
Melting Point 16.0°C to 18.0°C
Color Yellow
Packaging Glass bottle
Density 1.1200g/mL
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3COCH2COCO2C2H5
Beilstein 03, 747
Formula Weight 158.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl 2,4-dioxovalerate
Show More Show Less

EINECSNumber : 210-447-5