Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-amino-4-thiazoleacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass Bottle
Plastic bottle
53266-94-7
C7H10N2O2S
186.23
MFCD00005330
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
104454
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
CCOC(=O)CC1=CSC(=N1)N
53266-94-7
C7H10N2O2S
186.23
MFCD00005330
SHQNGLYXRFCPGZ-UHFFFAOYSA-N
ethyl 2-amino-4-thiazoleacetate, ethyl 2-2-aminothiazol-4-yl acetate, ethyl 2-2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazole-4-acetate, ethyl 2-amino-1,3-thiazol-4-yl acetate, ethyl 2-aminothiazol-4-acetate, 2-amino-4-thiazolyl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-amino-, ethyl ester, ethyl 2-amino-4-thiazolyl acetate
104454
ethyl 2-(2-amino-1,3-thiazol-4-yl)acetate
CCOC(=O)CC1=CSC(=N1)N
Color Yellow to Beige
Flash Point 185°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 5g
Melting Point 91.0°C to 95.0°C
Assay Percent Range 99%
Beilstein 27,336
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 186.23
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material Ethyl 2-amino-4-thiazoleacetate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 258-452-1