Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-butynoate, 99%, Acros Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
4341-76-8
C6H8O2
112.13
MFCD00015182
FCJJZKCJURDYNF-UHFFFAOYSA-N
ethyl 2-butynoate,ethyl tetrolate,2-butynoic acid ethyl ester,but-2-ynoic acid ethyl ester,2-butynoic acid, ethyl ester,tetrolic acid ethyl ester,ethyl-2-butynoate,ethyl 3-methylpropiolate,acmc-1aeo2,ethyl 1-propynecarboxylate
78043
ethyl but-2-ynoate
CCOC(=O)C#CC
4341-76-8
C6H8O2
112.13
MFCD00015182
FCJJZKCJURDYNF-UHFFFAOYSA-N
ethyl 2-butynoate,ethyl tetrolate,2-butynoic acid ethyl ester,but-2-ynoic acid ethyl ester,2-butynoic acid, ethyl ester,tetrolic acid ethyl ester,ethyl-2-butynoate,ethyl 3-methylpropiolate,acmc-1aeo2,ethyl 1-propynecarboxylate
78043
ethyl but-2-ynoate
CCOC(=O)C#CC
Flash Point 58°C
Quantity 10g
Boiling Point 160.0°C to 164.0°C
Color Brown to Gray
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Formula Weight 112.13
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Ethyl 2-butynoate
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning

EINECSNumber : 224-395-6