Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-fluoroacetoacetate, 97%, Acros Organics

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
1522-41-4
C6H9FO3
148.13
MFCD00215830
SHTFQLHOTAJQRJ-UHFFFAOYSA-N
ethyl 2-fluoroacetoacetate,ethyl2-fluoroacetoacetate,2-fluoro-3-oxo-butanoic acid ethyl ester,2-fluoroacetoacetic acid ethyl ester,ethyl 2-fluoro-3-oxobutyrate,2-fluoro-3-oxo-butyric acid ethyl ester,ethyl 2-fluoro-3-oxo-butanoate,ethyl-2-fluoroacetoacetate,ethyl-2-fluoro-3-oxobutanoate,fluoroacetoacetic acid ethyl ester
230078
ethyl 2-fluoro-3-oxobutanoate
CCOC(=O)C(C(=O)C)F
1522-41-4
C6H9FO3
148.13
MFCD00215830
SHTFQLHOTAJQRJ-UHFFFAOYSA-N
ethyl 2-fluoroacetoacetate,ethyl2-fluoroacetoacetate,2-fluoro-3-oxo-butanoic acid ethyl ester,2-fluoroacetoacetic acid ethyl ester,ethyl 2-fluoro-3-oxobutyrate,2-fluoro-3-oxo-butyric acid ethyl ester,ethyl 2-fluoro-3-oxo-butanoate,ethyl-2-fluoroacetoacetate,ethyl-2-fluoro-3-oxobutanoate,fluoroacetoacetic acid ethyl ester
230078
ethyl 2-fluoro-3-oxobutanoate
CCOC(=O)C(C(=O)C)F
Density 1.1810g/mL
Boiling Point 183°C
Flash Point 90°C
Specific Gravity 1.181
Color Colorless to Yellow
Infrared Spectrum Authentic
Quantity 25mL
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Linear Formula CH3COCH(F)COOCH2CH3
Solubility Information Solubility in water: soluble
Formula Weight 148.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 2-fluoroacetoacetate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Danger