Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-(methylsulfonyl)acetate, 97%, Thermo Scientific™

Catalog Number ac443200050
Click to view available options
Quantity:
5 g
4455-15-6
C5H10O4S
166.2
OCCWQCYBCZADCE-UHFFFAOYSA-N
ethyl methanesulfonylacetate, ethyl 2-methylsulfonyl acetate, ethyl methylsulfonylacetate, ethyl methylsulphonyl acetate, ethyl methanesulphonylacetate, ethyl methylsulfonyl acetate, ethyl 2-methanesulfonylacetate, acetic acid, methylsulfonyl-, ethyl ester, ethylmethanesulfonylacetate
78201
ethyl 2-methylsulfonylacetate
CCOC(=O)CS(=O)(=O)C
4455-15-6
C5H10O4S
166.2
OCCWQCYBCZADCE-UHFFFAOYSA-N
ethyl methanesulfonylacetate, ethyl 2-methylsulfonyl acetate, ethyl methylsulfonylacetate, ethyl methylsulphonyl acetate, ethyl methanesulphonylacetate, ethyl methylsulfonyl acetate, ethyl 2-methanesulfonylacetate, acetic acid, methylsulfonyl-, ethyl ester, ethylmethanesulfonylacetate
78201
ethyl 2-methylsulfonylacetate
CCOC(=O)CS(=O)(=O)C
Color Colorless to Yellow
Density 1.2590g/mL
Elemental Analysis 34.69 to 37.86% C, 5.82 to 6.31% H
Boiling Point 111°C to 113°C (0.2 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.259
Formula Weight 166.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-(methylsulfonyl)acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-702-3

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit