Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-nitrobenzoylacetate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
52119-39-8
C11H11NO5
237.21
MFCD03424821
OWZNCVIBJQPNEF-UHFFFAOYSA-N
ethyl 3-2-nitrophenyl-3-oxopropanoate,ethyl 2-nitrobenzoylacetate,acmc-1awhb,ethyl 3-2-nitrophenyl-3-oxo-propanoate,ethyl o-nitrobenzoylacetate,ethyl 2-o-nitrobenzoyl acetate,ethyl 2-2-nitrobenzoyl acetate,ethyl 2-o-nitrobenzoyl-acetate,ethyl 2-nitrobenzoyl acetate
10561837
ethyl 3-(2-nitrophenyl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-]
52119-39-8
C11H11NO5
237.21
MFCD03424821
OWZNCVIBJQPNEF-UHFFFAOYSA-N
ethyl 3-2-nitrophenyl-3-oxopropanoate,ethyl 2-nitrobenzoylacetate,acmc-1awhb,ethyl 3-2-nitrophenyl-3-oxo-propanoate,ethyl o-nitrobenzoylacetate,ethyl 2-o-nitrobenzoyl acetate,ethyl 2-2-nitrobenzoyl acetate,ethyl 2-o-nitrobenzoyl-acetate,ethyl 2-nitrobenzoyl acetate
10561837
ethyl 3-(2-nitrophenyl)-3-oxopropanoate
CCOC(=O)CC(=O)C1=CC=CC=C1[N+](=O)[O-]
Quantity 5g
Melting Point 35.0°C to 36.0°C
Infrared Spectrum Authentic
Color Orange to Brown
Assay Percent Range 96% min. (HPLC)
Formula Weight 237.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 2-nitrobenzoylacetate