Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle2-oxo-4-phenylbutyrate, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
64920-29-2
C12H14O3
206.241
STPXIOGYOLJXMZ-UHFFFAOYSA-N
ethyl 2-oxo-4-phenylbutyrate,ethyl benzylpyruvate,ethyl-2-oxo-4-phenylbutyrate,unii-5ynu9lpt51,5ynu9lpt51,2-oxo-4-phenylbutyric acid ethyl ester,2-oxo-4-phenyl butyric acid ethyl ester,ethyl 2-oxo-4-phenyl-butanoate,2-oxo-4-phenylbutanoic acid ethyl ester,ethyl2-oxo-4-phenylbutyrate
562087
ethyl 2-oxo-4-phenylbutanoate
CCOC(=O)C(=O)CCC1=CC=CC=C1
64920-29-2
C12H14O3
206.241
STPXIOGYOLJXMZ-UHFFFAOYSA-N
ethyl 2-oxo-4-phenylbutyrate,ethyl benzylpyruvate,ethyl-2-oxo-4-phenylbutyrate,unii-5ynu9lpt51,5ynu9lpt51,2-oxo-4-phenylbutyric acid ethyl ester,2-oxo-4-phenyl butyric acid ethyl ester,ethyl 2-oxo-4-phenyl-butanoate,2-oxo-4-phenylbutanoic acid ethyl ester,ethyl2-oxo-4-phenylbutyrate
562087
ethyl 2-oxo-4-phenylbutanoate
CCOC(=O)C(=O)CCC1=CC=CC=C1
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.084g/mL
Color Yellow
Assay Percent Range 95%
Linear Formula C6H5(CH)C(O)COOCHCH3
Formula Weight 206.24
Physical Form Liquid
Percent Purity ≥94.0%
Chemical Name or Material ethyle2-oxo-4-phenylbutyrate
Show More Show Less