Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Acros Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
Plastic bottle
886-86-2
C9H11NO2·CH4O3S
261.29
MFCD00013176
FQZJYWMRQDKBQN-UHFFFAOYSA-N
ethyl 3-aminobenzoate methanesulfonate,tricaine,tricaine methanesulfonate,finquel,metacaine,tricaine mesylate,3-aminobenzoic acid ethyl ester methanesulfonate,ethyl m-aminobenzoate methanesulfonate,ethyl 3-aminobenzoate methanesulfonic acid salt,3-aminobenzoic acid ethyl ester methanesulfonate salt
261501
ethyl 3-aminobenzoate;methanesulfonic acid
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O
886-86-2
C9H11NO2·CH4O3S
261.29
MFCD00013176
FQZJYWMRQDKBQN-UHFFFAOYSA-N
ethyl 3-aminobenzoate methanesulfonate,tricaine,tricaine methanesulfonate,finquel,metacaine,tricaine mesylate,3-aminobenzoic acid ethyl ester methanesulfonate,ethyl m-aminobenzoate methanesulfonate,ethyl 3-aminobenzoate methanesulfonic acid salt,3-aminobenzoic acid ethyl ester methanesulfonate salt
261501
ethyl 3-aminobenzoate;methanesulfonic acid
CCOC(=O)C1=CC(=CC=C1)N.CS(=O)(=O)O
Color White
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 146.0°C to 150.0°C
Assay Percent Range 98%
Linear Formula H2NC6H4CO2C2H5·CH3SO3H
Beilstein 14, 389
Merck Index 15, 9786
Formula Weight 261.29
Physical Form Fine Powder
Percent Purity 98%
Chemical Name or Material Ethyl 3-aminobenzoate, methanesulfonic acid salt
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.