Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chlorobutyrate, 97%, Acros Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
3153-36-4
C6H11ClO2
150.6
MFCD00001004
OPXNFHAILOHHFO-UHFFFAOYSA-N
ethyl 4-chlorobutyrate,butanoic acid, 4-chloro-, ethyl ester,ethyl gamma-chlorobutyrate,ethyl-4-chlorobutyrate,4-chloro-butyric acid ethyl ester,butyric acid, 4-chloro-, ethyl ester,ethyl .gamma.-chlorobutyrate,4-chlorobutyric acid ethyl ester,ethyl 4chlorobutyrate,acmc-1cqjw
18474
ethyl 4-chlorobutanoate
CCOC(=O)CCCCl
3153-36-4
C6H11ClO2
150.6
MFCD00001004
OPXNFHAILOHHFO-UHFFFAOYSA-N
ethyl 4-chlorobutyrate,butanoic acid, 4-chloro-, ethyl ester,ethyl gamma-chlorobutyrate,ethyl-4-chlorobutyrate,4-chloro-butyric acid ethyl ester,butyric acid, 4-chloro-, ethyl ester,ethyl .gamma.-chlorobutyrate,4-chlorobutyric acid ethyl ester,ethyl 4chlorobutyrate,acmc-1cqjw
18474
ethyl 4-chlorobutanoate
CCOC(=O)CCCCl
Refractive Index 1.4296 to 1.4316
Flash Point 51°C
Quantity 500g
Density 1.0700g/mL
Color Brown to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 186.0°C
Specific Gravity 1.07
Assay Percent Range 97%
Linear Formula Cl(CH2)3CO2C2H5
Beilstein 02, 278
Solubility Information Solubility in water: immiscible
Formula Weight 150.6
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 4-chlorobutyrate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning