Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle4-cyanobenzoate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
7153-22-2
C10H9NO2
175.19
JLSSWDFCYXSLQX-UHFFFAOYSA-N
benzoic acid, 4-cyano-, ethyl ester,4-cyanobenzoic acid ethyl ester,benzoic acid, 4-cyano-,ethyl ester,4-cyano-benzoic acid ethyl ester,ethyl p-cyanobenzoate,ethyl-4-cyanobenzoate,p-cyano ethyl benzoate,acmc-209ypo,ethyl 4-cyanobenzoate,4-cyanobenzoic acid, ethyl ester
81589
ethyl 4-cyanobenzoate
CCOC(=O)C1=CC=C(C=C1)C#N
7153-22-2
C10H9NO2
175.19
JLSSWDFCYXSLQX-UHFFFAOYSA-N
benzoic acid, 4-cyano-, ethyl ester,4-cyanobenzoic acid ethyl ester,benzoic acid, 4-cyano-,ethyl ester,4-cyano-benzoic acid ethyl ester,ethyl p-cyanobenzoate,ethyl-4-cyanobenzoate,p-cyano ethyl benzoate,acmc-209ypo,ethyl 4-cyanobenzoate,4-cyanobenzoic acid, ethyl ester
81589
ethyl 4-cyanobenzoate
CCOC(=O)C1=CC=C(C=C1)C#N
Melting Point 52°C to 55°C
Flash Point >110°C
Packaging Glass bottle
Quantity 5g
Assay Percent Range 99%
Linear Formula NCC6H4CO2C2H5
Beilstein 09, 846
Formula Weight 175.19
Chemical Name or Material Ethyl 4-cyanobenzoate, 99%

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning