Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle4-dimethylaminobenzoate, +99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
10287-53-3
C11H15NO2
193.25
MFCD00009115
FZUGPQWGEGAKET-UHFFFAOYSA-N
ethyl 4-dimethylamino benzoate,parbenate,benzoic acid, 4-dimethylamino-, ethyl ester,kayacure epa,ethyl-p-dimethylaminobenzoate,ethyl-4-dimethylaminobenzoate,n,n-dimethylbenzocaine,speedcure edb,ethyl p-dimethylaminobenzoate,unii-829s8d3y0x
25127
CHEBI:52073
ethyl 4-(dimethylamino)benzoate
CCOC(=O)C1=CC=C(C=C1)N(C)C
10287-53-3
C11H15NO2
193.25
MFCD00009115
FZUGPQWGEGAKET-UHFFFAOYSA-N
ethyl 4-dimethylamino benzoate,parbenate,benzoic acid, 4-dimethylamino-, ethyl ester,kayacure epa,ethyl-p-dimethylaminobenzoate,ethyl-4-dimethylaminobenzoate,n,n-dimethylbenzocaine,speedcure edb,ethyl p-dimethylaminobenzoate,unii-829s8d3y0x
25127
CHEBI:52073
ethyl 4-(dimethylamino)benzoate
CCOC(=O)C1=CC=C(C=C1)N(C)C
Specific Gravity 1.06
Melting Point 62°C to 65°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Color White to Yellow
Quantity 5g
Density 1.0600g/mL
Assay Percent Range 99+%
Linear Formula (CH3)2NC6H4CO2C2H5
Beilstein 14, I, 571
Solubility Information Solubility in water: insoluble. Other solubilities: >50g/100 mL acetone (20 c),18g/100mL ethanol (20 c),2g/100mL mineral oil (20 c)
Formula Weight 193.25
Physical Form Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material Ethyl 4-dimethylaminobenzoate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning