Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle4-fluorobenzoate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
451-46-7
C9H9FO2
168.17
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester,ethyl p-fluorobenzoate,benzoic acid, 4-fluoro-, ethyl ester,p-fluorobenzoic acid, ethyl ester,ethyl4-fluorobenzoate,4-fluoro-benzoic acid ethyl ester,benzoic acid, p-fluoro-, ethyl ester,ethyep-fluorobenzoate,pubchem3471,ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
451-46-7
C9H9FO2
168.17
MFCD00000351
UMPRJGKLMUDRHL-UHFFFAOYSA-N
4-fluorobenzoic acid ethyl ester,ethyl p-fluorobenzoate,benzoic acid, 4-fluoro-, ethyl ester,p-fluorobenzoic acid, ethyl ester,ethyl4-fluorobenzoate,4-fluoro-benzoic acid ethyl ester,benzoic acid, p-fluoro-, ethyl ester,ethyep-fluorobenzoate,pubchem3471,ethyl-4-fluorobenzoate
67976
ethyl 4-fluorobenzoate
CCOC(=O)C1=CC=C(C=C1)F
Density 1.1400g/mL
Refractive Index 1.4850 to 1.487
Melting Point 25°C to 27°C
Boiling Point 210°C
Flash Point 81°C
Specific Gravity 1.14
Packaging Glass bottle
Quantity 25g
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula FC6H4CO2C2H5
Beilstein 09, 334
Merck Index 10, 4076
Solubility Information Solubility in water: insoluble
Formula Weight 168.17
Physical Form Low Melting Solid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 4-fluorobenzoate
Show More Show Less