Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle4-hydroxybenzoate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
5kg
120-47-8
C9H10O3
166.18
MFCD00002353
NUVBSKCKDOMJSU-UHFFFAOYSA-N
ethylparaben,ethyl paraben,ethyl p-hydroxybenzoate,mycocten,ethyl parasept,4-hydroxybenzoic acid ethyl ester,easeptol,ethyl butex,ethyl parahydroxybenzoate,tegosept e
8434
CHEBI:86616
ethyl 4-hydroxybenzoate
CCOC(=O)C1=CC=C(C=C1)O
120-47-8
C9H10O3
166.18
MFCD00002353
NUVBSKCKDOMJSU-UHFFFAOYSA-N
ethylparaben,ethyl paraben,ethyl p-hydroxybenzoate,mycocten,ethyl parasept,4-hydroxybenzoic acid ethyl ester,easeptol,ethyl butex,ethyl parahydroxybenzoate,tegosept e
8434
CHEBI:86616
ethyl 4-hydroxybenzoate
CCOC(=O)C1=CC=C(C=C1)O
Boiling Point 297°C to 298°C
Melting Point 115°C to 118°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Color White
Quantity 100g
Assay Percent Range 99%
Linear Formula HOC6H4CO2C2H5
Beilstein 10, 159
Merck Index 15, 3890
Solubility Information Solubility in water: 1.7g/L (20°C). Other solubilities: freely soluble in alcohol and ether,soluble in carbon disulfide,slightly soluble in petroleum ether,chloroform
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Ethyl 4-hydroxybenzoate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.