Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-methyl-5-imidazolecarboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
51605-32-4
C7H10N2O2
154.17
MFCD00014487
VLDUBDZWWNLZCU-UHFFFAOYSA-N
ethyl 4-methyl-5-imidazolecarboxylate,ethyl 4-methyl-1h-imidazole-5-carboxylate,ethyl-4-methyl-5-imidazolecarboxylate,1h-imidazole-5-carboxylic acid, 4-methyl-, ethyl ester,1h-imidazole-4-carboxylic acid, 5-methyl-, ethyl ester,4-carboethoxy-5-methylimidazole,ethyl 4-methylimidazole-5-carboxylate,ethyl 5-methyl-4-imidazolecarboxylate,5-methyl-3h-imidazole-4-carboxylic acid ethyl ester,ethyl 5-methylimidazole-4-carboxylate
99200
ethyl 5-methyl-1H-imidazole-4-carboxylate
CCOC(=O)C1=C(NC=N1)C
51605-32-4
C7H10N2O2
154.17
MFCD00014487
VLDUBDZWWNLZCU-UHFFFAOYSA-N
ethyl 4-methyl-5-imidazolecarboxylate,ethyl 4-methyl-1h-imidazole-5-carboxylate,ethyl-4-methyl-5-imidazolecarboxylate,1h-imidazole-5-carboxylic acid, 4-methyl-, ethyl ester,1h-imidazole-4-carboxylic acid, 5-methyl-, ethyl ester,4-carboethoxy-5-methylimidazole,ethyl 4-methylimidazole-5-carboxylate,ethyl 5-methyl-4-imidazolecarboxylate,5-methyl-3h-imidazole-4-carboxylic acid ethyl ester,ethyl 5-methylimidazole-4-carboxylate
99200
ethyl 5-methyl-1H-imidazole-4-carboxylate
CCOC(=O)C1=C(NC=N1)C
Melting Point 204°C to 206°C
Quantity 50g
Color Tan to White
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Beilstein 25, I, 534
Solubility Information Solubility in water: insoluble in water
Formula Weight 154.17
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material Ethyl 4-methyl-5-imidazolecarboxylate, 98%
Show More Show Less

EINECSNumber : 257-315-3