Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-oxocyclohexanecarboxylate, 97%, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass Bottle
17159-79-4
C9H14O3
170.21
MFCD00013285
ZXYAWONOWHSQRU-UHFFFAOYSA-N
ethyl 4-oxocyclohexanecarboxylate,ethyl 4-cyclohexanonecarboxylate,4-ethoxycarbonyl cyclohexanone,ethyl cyclohexanone-4-carboxylate,ethyl-4-oxo-cyclohexane carboxylate,4-oxo-cyclohexanecarboxylic acid ethyl ester,cyclohexanecarboxylic acid, 4-oxo-, ethyl ester,ethyl4-oxocyclohexanecarboxylate,4-cyclohexanonecarboxylic acid ethyl ester,ethyl 4-oxocyclohexane carboxylate
317638
ethyl 4-oxocyclohexane-1-carboxylate
CCOC(=O)C1CCC(=O)CC1
17159-79-4
C9H14O3
170.21
MFCD00013285
ZXYAWONOWHSQRU-UHFFFAOYSA-N
ethyl 4-oxocyclohexanecarboxylate,ethyl 4-cyclohexanonecarboxylate,4-ethoxycarbonyl cyclohexanone,ethyl cyclohexanone-4-carboxylate,ethyl-4-oxo-cyclohexane carboxylate,4-oxo-cyclohexanecarboxylic acid ethyl ester,cyclohexanecarboxylic acid, 4-oxo-, ethyl ester,ethyl4-oxocyclohexanecarboxylate,4-cyclohexanonecarboxylic acid ethyl ester,ethyl 4-oxocyclohexane carboxylate
317638
ethyl 4-oxocyclohexane-1-carboxylate
CCOC(=O)C1CCC(=O)CC1
Flash Point 110°C
Boiling Point 150°C to 152°C (40.0mmHg)
Specific Gravity 1.076
Packaging Glass Bottle
Density 1.0760g/mL
Refractive Index 1.4580 to 1.461
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 250mg
Assay Percent Range 96% min. (GC)
Linear Formula (O=)C6H9CO2C2H5
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl 4-oxocyclohexanecarboxylate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning