Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle5-bromonicotinate, 99%, Acros Organics

Click to view available options
Quantity:
1g
20986-40-7
C8H8BrNO2
230.061
MFCD00040366
PCPIANOJERKFJI-UHFFFAOYSA-N
ethyl 5-bromonicotinate,5-bromonicotinic acid ethyl ester,ethyl 5-bromo-3-pyridinecarboxylate,3-pyridinecarboxylic acid, 5-bromo-, ethyl ester,5-bromo-3-pyridinecarboxylic acid ethyl ester,ethyl5-bromonicotinate,5-bromo nicotinic acid ethyl ester,5-bromo-nicotinic acid ethyl ester,ethyl 3-bromo-5-pyridinecarboxylate,3-bromo-5-pyridine-carboxylic acid ethyl ester
140785
ethyl 5-bromopyridine-3-carboxylate
CCOC(=O)C1=CC(=CN=C1)Br
20986-40-7
C8H8BrNO2
230.061
MFCD00040366
PCPIANOJERKFJI-UHFFFAOYSA-N
ethyl 5-bromonicotinate,5-bromonicotinic acid ethyl ester,ethyl 5-bromo-3-pyridinecarboxylate,3-pyridinecarboxylic acid, 5-bromo-, ethyl ester,5-bromo-3-pyridinecarboxylic acid ethyl ester,ethyl5-bromonicotinate,5-bromo nicotinic acid ethyl ester,5-bromo-nicotinic acid ethyl ester,ethyl 3-bromo-5-pyridinecarboxylate,3-bromo-5-pyridine-carboxylic acid ethyl ester
140785
ethyl 5-bromopyridine-3-carboxylate
CCOC(=O)C1=CC(=CN=C1)Br
Infrared Spectrum Authentic
Color White
Melting Point 39°C to 42°C
Quantity 1g
Assay Percent Range 99%
Formula Weight 230.06
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material ethyle5-bromonicotinate, 99%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.