Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-(chloromethyl)-2-furancarboxylate, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2528-00-9
C8H9ClO3
188.61
MFCD00173837
JBACYJRMCXLIQU-UHFFFAOYSA-N
ethyl 5-chloromethyl-2-furoate,ethyl 5-chloromethyl furan-2-carboxylate,ethyl 5-chloromethyl-2-furancarboxylate,5-chloromethyl-2-furancarboxylic acid ethyl ester,2-furancarboxylic acid, 5-chloromethyl-, ethyl ester,buttpark 9650-29,acmc-1cej4,ethyl 2-chloromethyl-5-furoate
263166
ethyl 5-(chloromethyl)furan-2-carboxylate
CCOC(=O)C1=CC=C(O1)CCl
2528-00-9
C8H9ClO3
188.61
MFCD00173837
JBACYJRMCXLIQU-UHFFFAOYSA-N
ethyl 5-chloromethyl-2-furoate,ethyl 5-chloromethyl furan-2-carboxylate,ethyl 5-chloromethyl-2-furancarboxylate,5-chloromethyl-2-furancarboxylic acid ethyl ester,2-furancarboxylic acid, 5-chloromethyl-, ethyl ester,buttpark 9650-29,acmc-1cej4,ethyl 2-chloromethyl-5-furoate
263166
ethyl 5-(chloromethyl)furan-2-carboxylate
CCOC(=O)C1=CC=C(O1)CCl
Quantity 5g
Boiling Point 115.0°C
Infrared Spectrum Authentic
Specific Gravity 1.225
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Color Yellow
Flash Point >110°C
Density 1.2250g/mL
Assay Percent Range 94% min. (GC)
Formula Weight 188.61
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Ethyl 5-(chloromethyl)-2-furancarboxylate
Show More Show Less

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning