Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-chloropyridazine-3-carboxylate, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
75680-92-1
C7H7ClN2O2
186.6
MFCD09908168
GVSVPKDEHFOXSW-UHFFFAOYSA-N
ethyl 6-chloro-3-pyridazinecarboxylate,6-chloro-pyridazine-3-carboxylic acid ethyl ester,3-pyridazinecarboxylic acid, 6-chloro-, ethyl ester,6-chloropyridazine-3-carboxylic acid ethyl ester,6-chloro-3-pyridazinecarboxylic acid ethyl ester,pubchem17694,acmc-1bd9t,6-chloro-pyridazine-3-carboxylicacidethylester,ksc641q6n,ethyl 3-chloropyridazine-6-carboxylate
10352425
ethyl 6-chloropyridazine-3-carboxylate
CCOC(=O)C1=NN=C(C=C1)Cl
75680-92-1
C7H7ClN2O2
186.6
MFCD09908168
GVSVPKDEHFOXSW-UHFFFAOYSA-N
ethyl 6-chloro-3-pyridazinecarboxylate,6-chloro-pyridazine-3-carboxylic acid ethyl ester,3-pyridazinecarboxylic acid, 6-chloro-, ethyl ester,6-chloropyridazine-3-carboxylic acid ethyl ester,6-chloro-3-pyridazinecarboxylic acid ethyl ester,pubchem17694,acmc-1bd9t,6-chloro-pyridazine-3-carboxylicacidethylester,ksc641q6n,ethyl 3-chloropyridazine-6-carboxylate
10352425
ethyl 6-chloropyridazine-3-carboxylate
CCOC(=O)C1=NN=C(C=C1)Cl
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 149.0°C to 152.0°C
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 186.6
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material Ethyl 6-chloropyridazine-3-carboxylate

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning