Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 7-methoxybenzofuran-2-carboxylate, 97%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
50551-58-1
C12H12O4
220.23
MFCD00060510
SXCMZYVXJUJIDJ-UHFFFAOYSA-N
ethyl 7-methoxybenzofuran-2-carboxylate,7-methoxybenzofuran-2-carboxylic acid ethyl ester,7-methoxybenzofuran-2-carboxylic acid, ethyl ester,ethyl 7-methoxybenzo b furan-2-carboxylate,7-methoxybenzofuran-2-carboxylic acidethyl ester,ethyl7-methoxybenzofuran-2-carboxylate,acmc-1aowz,2-ethoxycarbonyl-7-methoxybenzofuran,ethyl 4-methoxylbenzofunan-2-carboxylate,ethyl 7-methoxybenzo d furan-2-carboxylate
521284
ethyl 7-methoxy-1-benzofuran-2-carboxylate
CCOC(=O)C1=CC2=C(O1)C(=CC=C2)OC
50551-58-1
C12H12O4
220.23
MFCD00060510
SXCMZYVXJUJIDJ-UHFFFAOYSA-N
ethyl 7-methoxybenzofuran-2-carboxylate,7-methoxybenzofuran-2-carboxylic acid ethyl ester,7-methoxybenzofuran-2-carboxylic acid, ethyl ester,ethyl 7-methoxybenzo b furan-2-carboxylate,7-methoxybenzofuran-2-carboxylic acidethyl ester,ethyl7-methoxybenzofuran-2-carboxylate,acmc-1aowz,2-ethoxycarbonyl-7-methoxybenzofuran,ethyl 4-methoxylbenzofunan-2-carboxylate,ethyl 7-methoxybenzo d furan-2-carboxylate
521284
ethyl 7-methoxy-1-benzofuran-2-carboxylate
CCOC(=O)C1=CC2=C(O1)C(=CC=C2)OC
Melting Point 85°C to 90°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Yellow
Assay Percent Range 96% min. (GC)
Formula Weight 220.23
Physical Form Crystals
Percent Purity 97%
Chemical Name or Material Ethyl 7-methoxybenzofuran-2-carboxylate, 97%