Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl benzoylacetate, 90%, Acros Organics
Click to view available options
Quantity:
5g
50g
100g
500g
Packaging:
Glass bottle
Specifications
Boiling Point | 265°C to 270°C |
Flash Point | 140°C |
Specific Gravity | 1.106 |
Refractive Index | 1.5240 to 1.531 |
Density | 1.1060g/mL |
Color | Brown |
Quantity | 500g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Assay Percent Range | 88% min. (HPLC) |
Show More |
Safety and Handling
EINECSNumber : 202-295-3