Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl bromopyruvate, 80-85%, Acros Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
70-23-5
C5H7BrO3
195.01
MFCD00000204
VICYTAYPKBLQFB-UHFFFAOYSA-N
ethyl bromopyruvate,ethyl 3-bromopyruvate,propanoic acid, 3-bromo-2-oxo-, ethyl ester,bromopyruvic acid ethyl ester,ethyl bromo pyruvate,ethyl 3-bromo-2-oxopropionate,pyruvic acid, bromo-, ethyl ester,.beta.-bromopyruvic acid ethyl ester,3-bromo-2-oxo-propionic acid ethyl ester,beta-bromopyruvic acid ethyl ester
66144
ethyl 3-bromo-2-oxopropanoate
CCOC(=O)C(=O)CBr
70-23-5
C5H7BrO3
195.01
MFCD00000204
VICYTAYPKBLQFB-UHFFFAOYSA-N
ethyl bromopyruvate,ethyl 3-bromopyruvate,propanoic acid, 3-bromo-2-oxo-, ethyl ester,bromopyruvic acid ethyl ester,ethyl bromo pyruvate,ethyl 3-bromo-2-oxopropionate,pyruvic acid, bromo-, ethyl ester,.beta.-bromopyruvic acid ethyl ester,3-bromo-2-oxo-propionic acid ethyl ester,beta-bromopyruvic acid ethyl ester
66144
ethyl 3-bromo-2-oxopropanoate
CCOC(=O)C(=O)CBr
Color Pink-Red to Yellow
Boiling Point 100.0°C to 102.0°C (15.0 mmHg)
Quantity 100mL
Density 1.5500g/mL
Infrared Spectrum Authentic
Flash Point 98°C
Refractive Index 1.4675 to 1.4765
Packaging Glass bottle
Specific Gravity 1.55
Assay Percent Range 80-85%
Linear Formula BrCH2COCO2C2H5
Beilstein 03, II, 409
Formula Weight 195.01
Physical Form Liquid
Percent Purity 80-85%
Chemical Name or Material Ethyl bromopyruvate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger