Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl chloroacetate, 99%, Acros Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
105-39-5
C4H7ClO2
122.55
MFCD00000932
VEUUMBGHMNQHGO-UHFFFAOYSA-N
ethyl chloroacetate,ethyl chloracetate,ethyl monochloroacetate,acetic acid, chloro-, ethyl ester,chloroacetic acid ethyl ester,ethyl monochloracetate,ethyl chloroethanoate,ethyl alpha-chloroacetate,2-chloroacetic acid ethyl ester,unii-6h07teh4tf
7751
ethyl 2-chloroacetate
CCOC(=O)CCl
105-39-5
C4H7ClO2
122.55
MFCD00000932
VEUUMBGHMNQHGO-UHFFFAOYSA-N
ethyl chloroacetate,ethyl chloracetate,ethyl monochloroacetate,acetic acid, chloro-, ethyl ester,chloroacetic acid ethyl ester,ethyl monochloracetate,ethyl chloroethanoate,ethyl alpha-chloroacetate,2-chloroacetic acid ethyl ester,unii-6h07teh4tf
7751
ethyl 2-chloroacetate
CCOC(=O)CCl
Density 1.1500g/mL
Viscosity 1.27 mPa.s (20°C)
Infrared Spectrum Authentic
Melting Point -26.0°C
Boiling Point 142.0°C to 145.0°C
Packaging Glass bottle
Flash Point 53°C
Specific Gravity 1.15
Refractive Index 1.4200 to 1.4220
Quantity 250mL
Color Colorless
Assay Percent Range 99%
Linear Formula ClCH2CO2C2H5
Beilstein 02,197
Fieser 13,348
Merck Index 15,3838
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Formula Weight 122.55
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethyl chloroacetate
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger