Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl chloroformate, 99%, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
541-41-3
C3H5ClO2
108.52
MFCD00000644
RIFGWPKJUGCATF-UHFFFAOYSA-N
ethyl chloroformate,ethyl chlorocarbonate,cathyl chloride,ethoxycarbonyl chloride,chloroformic acid ethyl ester,ethylchloroformate,carbonochloridic acid, ethyl ester,chloro ethoxy methanone,ethylchloorformiaat,etil cloroformiato
10928
ethyl carbonochloridate
CCOC(=O)Cl
541-41-3
C3H5ClO2
108.52
MFCD00000644
RIFGWPKJUGCATF-UHFFFAOYSA-N
ethyl chloroformate,ethyl chlorocarbonate,cathyl chloride,ethoxycarbonyl chloride,chloroformic acid ethyl ester,ethylchloroformate,carbonochloridic acid, ethyl ester,chloro ethoxy methanone,ethylchloorformiaat,etil cloroformiato
10928
ethyl carbonochloridate
CCOC(=O)Cl
Coupling reagent
Viscosity 0.46 mPa.s (20°C)
Quantity 800mL
Infrared Spectrum Authentic
Color White
Flash Point 10°C
Boiling Point 94.0°C
Density 1.1350g/mL
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3950 to 1.3960
Specific Gravity 1.135
Melting Point -81.0°C
Assay Percent Range 1% max. Diethyl carbonate (GC)
Linear Formula ClCO2C2H5
Merck Index 15,3839
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol, soluble in benzene, chloroform and ether, soluble in most organic solvents
Formula Weight 108.52
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material Ethyl chloroformate
Show More Show Less

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a POISON CENTER or doctor/physi

GHS Signal Word: Danger

EINECSNumber : 208-778-5