Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methanesulfonate, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
500g
Packaging:
Glass bottle
62-50-0
C3H8O3S
124.15
MFCD00007559
PLUBXMRUUVWRLT-UHFFFAOYSA-N
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
6113
CHEBI:23994
ethyl methanesulfonate
CCOS(=O)(=O)C
62-50-0
C3H8O3S
124.15
MFCD00007559
PLUBXMRUUVWRLT-UHFFFAOYSA-N
ethyl mesylate, ethyl methanesulphonate, half-myleran, methanesulfonic acid, ethyl ester, methanesulfonic acid ethyl ester, methylsulfonic acid, ethyl ester, rcra waste number u119, ethyl methansulphonate, ethyl ester of methylsulfonic acid, ethyl ester of methanesulfonic acid
6113
CHEBI:23994
ethyl methanesulfonate
CCOS(=O)(=O)C
Flash Point 100°C
Density 1.1600g/mL
Boiling Point 213.0°C to 214.0°C
Packaging Glass bottle
Quantity 500g
Refractive Index 1.4170 to 1.4190
Specific Gravity 1.16
Assay Percent Range 99%
Linear Formula CH3SO3C2H5
Beilstein 04,5
Merck Index 15,3882
Solubility Information Solubility in water: 50-100g/L (27°C)
Formula Weight 124.15
Percent Purity 99%
Chemical Name or Material Ethyl methanesulfonate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause cancer.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-536-7