Alfa Aesar™ Ethylene 1,2-dithiocyanate, 98%

Manufacturer: Alfa Aesar™ B2218214

Quantity 25g

 View more versions of this product

Catalog No. AAB2218214

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 629-17-4
    Molecular Formula NCSCH2CH2SCN
    Formula Weight 144.21
    MDL Number MFCD00013807
    UN Number UN2811
    Synonym 1,2-Bis(thiocyanato)ethane
    Chemical Name or Material Ethylene 1,2-dithiocyanate

    Specifications

    Formula Weight 144.21
    Quantity 25g
    TSCA TSCA
    Molecular Formula NCSCH2CH2SCN
    MDL Number MFCD00013807
    CAS 629-17-4
    UN Number UN2811
    Synonym 1,2-Bis(thiocyanato)ethane
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 88° to 92°C
    Odor Irritating
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number XK9862500
    Chemical Name or Material Ethylene 1,2-dithiocyanate
    Assay 98%

    GHS H Statement
    H300-H315-H319-H335
    Fatal if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit