Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethylene 1,2-dithiocyanate, 98%

Click to view available options
Quantity:
5g
629-17-4
C4H4N2S2
144.21
MFCD00013807
LILPOSVDSDEBCD-UHFFFAOYSA-N
ethylene 1,2-dithiocyanate,ethylene dithiocyanate,ethylene thiocyanate,ethylenedithiocyanate,1,2-dithiocyanatoethane,1,2-bis thiocyanato ethane,1,2-dithiocyanoethane,1,2-ethanediyl thiocyanate,thiocyanic acid, 1,2-ethanediyl ester,thiocyanic acid, c,c'-1,2-ethanediyl ester
69416
2-thiocyanatoethyl thiocyanate
C(CSC#N)SC#N
629-17-4
C4H4N2S2
144.21
MFCD00013807
LILPOSVDSDEBCD-UHFFFAOYSA-N
ethylene 1,2-dithiocyanate,ethylene dithiocyanate,ethylene thiocyanate,ethylenedithiocyanate,1,2-dithiocyanatoethane,1,2-bis thiocyanato ethane,1,2-dithiocyanoethane,1,2-ethanediyl thiocyanate,thiocyanic acid, 1,2-ethanediyl ester,thiocyanic acid, c,c'-1,2-ethanediyl ester
69416
2-thiocyanatoethyl thiocyanate
C(CSC#N)SC#N
Quantity 5g
Melting Point 88°C to 92°C
Odor Irritating
UN Number UN2811
Formula Weight 144.21
Percent Purity 98%
Chemical Name or Material Ethylene 1,2-dithiocyanate

GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XK9862500

TSCA : TSCA