Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene/acrylic acid copolymer, Acros Organics

Click to view available options
Quantity:
100g
250g
9010-77-9
C5H8O2
100.117
MFCD00133995
QHZOMAXECYYXGP-UHFFFAOYSA-N
62701
ethene;prop-2-enoic acid
C=C.C=CC(=O)O
9010-77-9
C5H8O2
100.117
MFCD00133995
QHZOMAXECYYXGP-UHFFFAOYSA-N
62701
ethene;prop-2-enoic acid
C=C.C=CC(=O)O
Linear Formula (C3H4O2·C2H4)x
Packaging Glass bottle
Quantity 100g
Chemical Name or Material Ethylene/acrylic acid copolymer