Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene diacrylate, 95%, stabilized, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
2274-11-5
C8H10O4
170.17
MFCD00008629
KUDUQBURMYMBIJ-UHFFFAOYSA-N
ethylene diacrylate,ethylene acrylate,ethylene glycol diacrylate,acrylic acid, ethylene ester,acrylic acid, ethylene glycol diester,2-propenoic acid, 1,2-ethanediyl ester,acrylic acid, diester with ethylene glycol,1,2-ethanediyl diacrylate,ethylene glycol dipropenoate,unii-42q902riye
75282
2-prop-2-enoyloxyethyl prop-2-enoate
C=CC(=O)OCCOC(=O)C=C
2274-11-5
C8H10O4
170.17
MFCD00008629
KUDUQBURMYMBIJ-UHFFFAOYSA-N
ethylene diacrylate,ethylene acrylate,ethylene glycol diacrylate,acrylic acid, ethylene ester,acrylic acid, ethylene glycol diester,2-propenoic acid, 1,2-ethanediyl ester,acrylic acid, diester with ethylene glycol,1,2-ethanediyl diacrylate,ethylene glycol dipropenoate,unii-42q902riye
75282
2-prop-2-enoyloxyethyl prop-2-enoate
C=CC(=O)OCCOC(=O)C=C
Flash Point 101°C
Stabilizer 100ppm MEHQ
Specific Gravity 1.04
Packaging Glass bottle
Refractive Index 1.4520 to 1.4560
Quantity 25mL
Color Colorless to Yellow
Boiling Point 62.0°C to 65.0°C (2.0mmHg)
Density 1.0400g/mL
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Beilstein 02, 1469
Solubility Information Solubility in water: slightly soluble
Formula Weight 170.17
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Ethylene diacrylate, 90%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lastin

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

EINECSNumber : 218-886-4