Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, ≥99%, Spectrophotometric Grade, Honeywell Riedel-de Haën™

Catalog Number p-7120430
Click to view available options
Quantity:
1 L
2 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
107-21-1
C2H6O2
62.068
MFCD00002885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
107-21-1
C2H6O2
62.068
MFCD00002885
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

Spectrophotometric Grade,≥99%

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
TRUSTED_SUSTAINABILITY
Melting Point -12°C
Linear Formula HOCH2CH2OH
UN Number UN3082
Beilstein 505945
Formula Weight 62.07g/mol
Percent Purity ≥99%
Grade Spectrophotometric
Density 1.113g/mL at 25°C
Boiling Point 195°C to 198°C
Quantity 1 L
Explosion Limit 0.153
Flash Point 111°C (231.8°F)
Ignition Point 752°F
Packaging Glass Bottle
Vapor Pressure 0.08mmHg (20°C)
Chemical Name or Material Ethylene Glycol
Show More Show Less
Hazard Statement P260-P301 + P312 + P330
Precautionary Statement H302-H373
DOTInformation 9/PGIII
EINECSNumber 203-473-3
RTECSNumber KW2975000
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit