Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

107-21-1
C2H6O2
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
174
CHEBI:30742

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
CAS 107-21-1
Molecular Formula C2H6O2
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES C(CO)O
IUPAC Name ethane-1,2-diol
Molecular Weight (g/mol) 62.068
PubChem CID 174
ChEBI CHEBI:30742
Formula Weight 62.07
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Flash Point 111.0°C (231.8°F)
Quantity 4L
Chemical Name or Material Ethylene Glycol
Grade BAKER ANALYZED™ Reagent
Show More Show Less