Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
107-21-1
C2H6O2
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O
107-21-1
C2H6O2
62.068
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethylene glycol,1,2-ethanediol,glycol,monoethylene glycol,1,2-dihydroxyethane,2-hydroxyethanol,glycol alcohol,ethylene alcohol,fridex,tescol
174
CHEBI:30742
ethane-1,2-diol
C(CO)O

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 62.07
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 1.10kg
Flash Point 111.00°C
Quantity 4L
Chemical Name or Material Ethylene Glycol