Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol diethyl ether, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
Packaging:
Glass bottle
629-14-1
C6H14O2
118.18
MFCD00009253
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
ethylene glycol diethyl ether,diethyl cellosolve,ethane, 1,2-diethoxy,hisolve eme,3,6-dioxaoctane,glyme-1,diethyl glycol,2-ethoxyethyl ethyl ether,egdee,diethylether ethylenglykolu
12375
1,2-diethoxyethane
CCOCCOCC
629-14-1
C6H14O2
118.18
MFCD00009253
LZDKZFUFMNSQCJ-UHFFFAOYSA-N
ethylene glycol diethyl ether,diethyl cellosolve,ethane, 1,2-diethoxy,hisolve eme,3,6-dioxaoctane,glyme-1,diethyl glycol,2-ethoxyethyl ethyl ether,egdee,diethylether ethylenglykolu
12375
1,2-diethoxyethane
CCOCCOCC
Viscosity 0.7 mPa.s (20°C)
Water 0.1% max.
Melting Point -74.0°C
Quantity 2.5L
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.3910 to 1.3930
Boiling Point 121.0°C
Density 0.8417g/mL
Packaging Glass bottle
Flash Point 19°C
Specific Gravity 0.8417
Assay Percent Range 98% min. (GC)
Linear Formula C2H5OCH2CH2OC2H5
Beilstein 01, 468
Solubility Information Solubility in water: 20.4%.
Formula Weight 118.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Ethylene glycol diethyl ether
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May damage the unborn child.
Suspected of damaging fertility.
Highly flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep container tightly closed.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES

GHS Signal Word: Danger

EINECSNumber : 211-076-1