Alfa Aesar™ Ethylene glycol dimethacrylate, 98%, stab. with 100ppm 4-methoxyphenol

Crosslinking monomer

Manufacturer: Alfa Aesar™ 44151A6

Quantity 4kg

 View more versions of this product

Catalog No. AA44151A6

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 97-90-5
    Molecular Formula C10H14O4
    Linear Formula H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
    Formula Weight 198.21
    MDL Number MFCD00008590
    Beilstein 1776663
    Synonym Ethylene dimethacrylate
    Chemical Name or Material Ethylene glycol dimethacrylate, Stabilized with 100ppm 4-methoxyphenol

    Specifications

    Formula Weight 198.21
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1776663
    Health Hazard 2 GHS H Statement
    H317-H335
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Density 1.051g/mL
    Boiling Point 83° to 85°C (1mmHg)
    Color Colorless to pale yellow
    Quantity 4kg
    RTECS Number OZ4400000
    Physical Form Liquid
    Linear Formula H2C=C(CH3)CO2CH2CH2OCOC(CH3)=CH2
    CAS 97-90-5
    Synonym Ethylene dimethacrylate
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    Molecular Formula C10H14O4
    MDL Number MFCD00008590
    Health Hazard 1 Warning
    Refractive Index 1.4540
    Flash Point >110°C (230°F)
    Chemical Name or Material Ethylene glycol dimethacrylate, Stabilized with 100ppm 4-methoxyphenol
    Assay 98%

    GHS H Statement
    H317-H335
    May cause an allergic skin reaction.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P304+P340-P312-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit