Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Iron Chloride Free, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
107-21-1
107-21-1
Packaging Amber Glass Bottle
Acidity 5e-05
Water 0.001
Residue after Ignition 5e-05
Quantity 500 mL
Specific Gravity 1.1151 - 1.1156
CAS Max % 1
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.