Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Reagent, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
107-21-1
C2H6O2
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
OCCO
107-21-1
C2H6O2
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
OCCO

E1051, 107-21-1, C2H6O2

Spectrum™ Ethylene Glycol, Reagent is a colorless, odorless, sweet tasting organic compound that has a syrupy texture and is a liquid. It has many lab applications including use as an antifreeze, use as a desiccant, protecting group for carbonyls during chemical synthesis and as a preservative for biological specimens. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
CAS Max % 1
Acidity 5e-005
Percent Purity 99%
Packaging Poly Bottle
Quantity 6 x 1 L