Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene trithiocarbonate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
822-38-8
C3H4S3
136.245
MFCD00005493
XCWPBWWTGHQKDR-UHFFFAOYSA-N
ethylene trithiocarbonate,cyclic ethylene trithiocarbonate,trithiocarbonic acid, cyclic ethylene ester,carbonic acid, trithio-, cyclic ethylene ester,cylic ethylene trithiocarbonate,acmc-209pnw,1,3-dithiolan-2-thione,5-19-04-00014 beilstein handbook reference,ksc449a8r
13196
1,3-dithiolane-2-thione
C1CSC(=S)S1
822-38-8
C3H4S3
136.245
MFCD00005493
XCWPBWWTGHQKDR-UHFFFAOYSA-N
ethylene trithiocarbonate,cyclic ethylene trithiocarbonate,trithiocarbonic acid, cyclic ethylene ester,carbonic acid, trithio-, cyclic ethylene ester,cylic ethylene trithiocarbonate,acmc-209pnw,1,3-dithiolan-2-thione,5-19-04-00014 beilstein handbook reference,ksc449a8r
13196
1,3-dithiolane-2-thione
C1CSC(=S)S1
Density 163g/mL
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 97%
Formula Weight 136.26
Physical Form Low Melting Solid
Percent Purity ≥96.0%
Chemical Name or Material Ethylene trithiocarbonate