Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(diphenylphosphino)ethane, 98+%, Acros Organics™

Click to view available options
Quantity:
1g
10g
50g
250g
Packaging:
Glass bottle
1663-45-2
C26H24P2
398.41
MFCD00003047
QFMZQPDHXULLKC-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane,diphos,ethylenebis diphenylphosphine,bis diphenylphosphine ethane,bis 1,2-diphenylphosphino ethane,phosphine, 1,2-ethanediylbis diphenyl,phosphine, ethylenebis diphenyl,1,2-bis diphenylphosphine ethane,1,2-bis diphenylphosphino-ethane,unii-kl33qe52i4
74267
CHEBI:30669
2-diphenylphosphanylethyl(diphenyl)phosphane
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
1663-45-2
C26H24P2
398.41
MFCD00003047
QFMZQPDHXULLKC-UHFFFAOYSA-N
1,2-bis diphenylphosphino ethane,diphos,ethylenebis diphenylphosphine,bis diphenylphosphine ethane,bis 1,2-diphenylphosphino ethane,phosphine, 1,2-ethanediylbis diphenyl,phosphine, ethylenebis diphenyl,1,2-bis diphenylphosphine ethane,1,2-bis diphenylphosphino-ethane,unii-kl33qe52i4
74267
CHEBI:30669
2-diphenylphosphanylethyl(diphenyl)phosphane
C1=CC=C(C=C1)P(CCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Melting Point 138.0°C to 142.0°C
Quantity 50g
Assay Percent Range 98+%
Linear Formula (C6H5)2PCH2CH2P(C6H5)2
Merck Index 15,1254
Formula Weight 398.41
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98%
Chemical Name or Material 1, 2-Bis(diphenylphosphino)ethane, 98+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 216-769-2

TSCA : TSCA