Alfa Aesar™ Ethylenediamine, 99%

Manufacturer: Alfa Aesar™ A12132DLS

Quantity 4 x 2.5L

 View more versions of this product

Catalog No. AAA12132DLS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-15-3
    Molecular Formula H2NCH2CH2NH2
    Linear Formula H2NCH2CH2NH2
    Formula Weight 60.1
    MDL Number MFCD00008204
    UN Number UN1604
    Beilstein 605263
    Merck Index 14,3795
    Synonym 1,2-Diaminoethane; 1,2-Ethanediamine
    Chemical Name or Material Ethylenediamine

    Specifications

    Formula Weight 60.1
    Boiling Point 117° to 118°C
    Melting Point 8° to 11°C
    Assay Percent Range 99%
    Molecular Formula H2NCH2CH2NH2
    Storage Note 1 Air Sensitive
    Health Hazard 2 GHS H Statement
    H334-H314-H226-H302-H312-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00008204
    UN Number UN1604
    Beilstein 605263
    Merck Index 14,3795
    Density 0.899g/mL
    Health Hazard 1 Danger
    Refractive Index 1.4565
    DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: ETHYLENEDIAMINE
    Sensitivity Air Sensitive
    Linear Formula H2NCH2CH2NH2
    Synonym 1,2-Diaminoethane; 1,2-Ethanediamine
    Solubility Information Soluble in water, alcohol; Slightly soluble in ether
    Odor Ammonia-like
    CAS 107-15-3
    RTECS Number KH8575000
    Color colorless to pale yellow
    Quantity 4 x 2.5L
    Chemical Name or Material Ethylenediamine
    Assay 99%

    Description

    Applications
    Ethylenediamine is a strongly basic amine useful as a building block in chemical synthesis. It is used as a solvent to dissolve proteins such as albumins and casein. It is widely used for color photography developers, binders, adhesives, fabric softeners, curing agents for epoxys and dyes. As a corrosion inhibitor, it plays a vital role in paints and coolants. It is used as an intermediate in the preparation of polyamide resins, fuel additives and lubricants. It acts as a precursor for many polymers like polyurethane fibers and poly(amidoamine), ethylenediamine dihydroiodide (EDDI) as well as the bleaching activator, tetraacetylethylenediamine. It is an important chelating ligand used in the preparation of coordination compounds viz. tris(ethylenediamine)cobalt(III) chloride. It is also involved in the manufacture of many industrial chemicals and forms derivatives with carboxylic acids, nitriles, alcohols, alkylating agents, carbon disulfide, aldehydes and ketones. It is a basic building block to prepare heterocyclic compound such as imidazolidines.

    Notes
    Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, phosphorus halides, aldehydes and organic halides.

    GHS H Statement
    H334-H314-H226-H302-H312-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Air Sensitive

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit